missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-2-butene, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 201510250

 View more versions of this product

Catalog No. AC201510250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-2-butene
13294-71-8
100.0
1.3200g/mL
<1°C
97.5% min. (GC)
C4H7Br
CH3CH=C(Br)CH3
25 mL
1.32
Solubility in water: immiscible
C\C=C(/C)Br
135.00
135
Liquid
98%
97.5
Brown to Yellow
82°C to 90°C (740.0 mmHg)
Authentic
Glass bottle
1.4590 to 1.462
MFCD00000141
01, 205
2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene #
UILZQFGKPHAAOU-ONEGZZNKSA-N
(E)-2-bromobut-2-ene
5364387
98%

Chemical Identifiers

13294-71-8
135.00
UILZQFGKPHAAOU-ONEGZZNKSA-N
5364387
C\C=C(/C)Br
C4H7Br
MFCD00000141
2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene #
(E)-2-bromobut-2-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-316-2

TSCA : TSCA

Recommended Storage : Product may darken during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only