missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4'-(1-pyrrolidinyl)acetophenone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1540706

 View more versions of this product

Catalog No. AAL1540706


Explore available promotions

Description

Description

2-Bromo-4'-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications

Specifications

2-Bromo-4'-(1-pyrrolidinyl)acetophenone
120°C to 122°C
MFCD01075690
UN3261
2-bromo-1-4-pyrrolidin-1-yl phenyl ethanone, alpha-bromo-4-1-pyrrolidino acetophenone, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl ethanone, ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl, 2-bromo-1-4-pyrrolidin-1-yl phenyl ethan-1-one, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethan-1-one, 4-pyrrolidin-1-yl phenacyl bromide, 2-bromo-1-4-pyrrolidinylphenyl ethan-1-one, acmc-209fm8
WLRIUCQUMKJOGT-UHFFFAOYSA-N
2-bromo-1-(4-pyrrolidin-1-ylphenyl)ethanone
3681785
97%
216144-18-2
C12H14BrNO
5 g
8549816
Insoluble in water.
C1CCN(C1)C2=CC=C(C=C2)C(=O)CBr
268.154
268.16

Chemical Identifiers

216144-18-2
268.154
WLRIUCQUMKJOGT-UHFFFAOYSA-N
3681785
C1CCN(C1)C2=CC=C(C=C2)C(=O)CBr
C12H14BrNO
MFCD01075690
2-bromo-1-4-pyrrolidin-1-yl phenyl ethanone, alpha-bromo-4-1-pyrrolidino acetophenone, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl ethanone, ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl, 2-bromo-1-4-pyrrolidin-1-yl phenyl ethan-1-one, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethan-1-one, 4-pyrrolidin-1-yl phenacyl bromide, 2-bromo-1-4-pyrrolidinylphenyl ethan-1-one, acmc-209fm8
2-bromo-1-(4-pyrrolidin-1-ylphenyl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only