Learn More
2-Bromo-4'-(1-pyrrolidinyl)acetophenone, 97%, Thermo Scientific Chemicals
$159.10 - $568.86
Chemical Identifiers
CAS | 216144-18-2 |
---|---|
Molecular Formula | C12H14BrNO |
Molecular Weight (g/mol) | 268.154 |
MDL Number | MFCD01075690 |
InChI Key | WLRIUCQUMKJOGT-UHFFFAOYSA-N |
Synonym | 2-bromo-1-4-pyrrolidin-1-yl phenyl ethanone, alpha-bromo-4-1-pyrrolidino acetophenone, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl ethanone, ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl, 2-bromo-1-4-pyrrolidin-1-yl phenyl ethan-1-one, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethan-1-one, 4-pyrrolidin-1-yl phenacyl bromide, 2-bromo-1-4-pyrrolidinylphenyl ethan-1-one, acmc-209fm8 |
PubChem CID | 3681785 |
IUPAC Name | 2-bromo-1-(4-pyrrolidin-1-ylphenyl)ethanone |
SMILES | C1CCN(C1)C2=CC=C(C=C2)C(=O)CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1540706
|
Thermo Scientific Chemicals
L1540706 |
5 g |
Each for $159.10
|
|
|||||
AAL1540714
|
Thermo Scientific Chemicals
L1540714 |
25 g |
Each for $568.86
|
|
|||||
Description
2-Bromo-4'-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4′-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Chemical Identifiers
216144-18-2 | |
268.154 | |
WLRIUCQUMKJOGT-UHFFFAOYSA-N | |
3681785 | |
C1CCN(C1)C2=CC=C(C=C2)C(=O)CBr |
C12H14BrNO | |
MFCD01075690 | |
2-bromo-1-4-pyrrolidin-1-yl phenyl ethanone, alpha-bromo-4-1-pyrrolidino acetophenone, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl ethanone, ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl, 2-bromo-1-4-pyrrolidin-1-yl phenyl ethan-1-one, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethan-1-one, 4-pyrrolidin-1-yl phenacyl bromide, 2-bromo-1-4-pyrrolidinylphenyl ethan-1-one, acmc-209fm8 | |
2-bromo-1-(4-pyrrolidin-1-ylphenyl)ethanone |
Specifications
216144-18-2 | |
C12H14BrNO | |
5 g | |
8549816 | |
Insoluble in water. | |
C1CCN(C1)C2=CC=C(C=C2)C(=O)CBr | |
268.154 | |
268.16 | |
2-Bromo-4'-(1-pyrrolidinyl)acetophenone |
120°C to 122°C | |
MFCD01075690 | |
UN3261 | |
2-bromo-1-4-pyrrolidin-1-yl phenyl ethanone, alpha-bromo-4-1-pyrrolidino acetophenone, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl ethanone, ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl, 2-bromo-1-4-pyrrolidin-1-yl phenyl ethan-1-one, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethan-1-one, 4-pyrrolidin-1-yl phenacyl bromide, 2-bromo-1-4-pyrrolidinylphenyl ethan-1-one, acmc-209fm8 | |
WLRIUCQUMKJOGT-UHFFFAOYSA-N | |
2-bromo-1-(4-pyrrolidin-1-ylphenyl)ethanone | |
3681785 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only