missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-6-pyrrolidin-1-ylpyridine, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC38010DA

 View more versions of this product

Catalog No. CC38010DA


Explore available promotions

Description

Description

Specifications

Specifications

2-Bromo-6-pyrrolidin-1-ylpyridine
89.5
90%
C9H11BrN2
1 g
HZOWJDJNPISFJC-UHFFFAOYSA-N
2-bromo-6-pyrrolidin-1-ylpyridine
15470277
90%
230618-41-4
100.0
Amber Glass Bottle
MFCD08271892
2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl
C1CCN(C1)C2=NC(=CC=C2)Br
227.105
227.1

Chemical Identifiers

230618-41-4
227.105
HZOWJDJNPISFJC-UHFFFAOYSA-N
15470277
C1CCN(C1)C2=NC(=CC=C2)Br
C9H11BrN2
MFCD08271892
2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl
2-bromo-6-pyrrolidin-1-ylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions