missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-6-pyrrolidin-1-ylpyridine, ≥97%, Thermo Scientific™

$82.63 - $82.63

Chemical Identifiers

CAS 230618-41-4
Molecular Formula C9H11BrN2
Molecular Weight (g/mol) 227.105
MDL Number MFCD08271892
InChI Key HZOWJDJNPISFJC-UHFFFAOYSA-N
Synonym 2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl
PubChem CID 15470277
IUPAC Name 2-bromo-6-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=NC(=CC=C2)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC38010DA
SDS View Documents
Thermo Scientific Chemicals
CC38010DA
1 g
Each for $82.63
 
Description

Description

Specifications

Chemical Identifiers

230618-41-4
227.105
HZOWJDJNPISFJC-UHFFFAOYSA-N
15470277
C1CCN(C1)C2=NC(=CC=C2)Br
C9H11BrN2
MFCD08271892
2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl
2-bromo-6-pyrrolidin-1-ylpyridine

Specifications

230618-41-4
100.0
Amber Glass Bottle
MFCD08271892
2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl
C1CCN(C1)C2=NC(=CC=C2)Br
227.105
227.1
2-Bromo-6-pyrrolidin-1-ylpyridine
89.5
90%
C9H11BrN2
1 g
HZOWJDJNPISFJC-UHFFFAOYSA-N
2-bromo-6-pyrrolidin-1-ylpyridine
15470277
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'