missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America B0552100G
Specifications
2-Bromobenzoic Acid | |
150°C | |
C7H5BrO2 | |
100 g | |
XRXMNWGCKISMOH-UHFFFAOYSA-N | |
2-bromobenzoic acid | |
6940 | |
≥98.0% (GC,T) |
88-65-3 | |
White-Yellow | |
MFCD00002402 | |
o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657 | |
C1=CC=C(C(=C1)C(=O)O)Br | |
201.019 | |
201.02 | |
Crystalline Powder |
Chemical Identifiers
88-65-3 | |
201.019 | |
XRXMNWGCKISMOH-UHFFFAOYSA-N | |
6940 | |
C1=CC=C(C(=C1)C(=O)O)Br |
C7H5BrO2 | |
MFCD00002402 | |
o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657 | |
2-bromobenzoic acid |
Safety and Handling
EINECSNumber : (3)-0062
RTECSNumber : DG4448035
TSCA : Yes