missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromobenzoic Acid 98.0+%, TCI America™
$29.41 - $223.19
Chemical Identifiers
CAS | 88-65-3 |
---|---|
Molecular Formula | C7H5BrO2 |
Molecular Weight (g/mol) | 201.019 |
MDL Number | MFCD00002402 |
InChI Key | XRXMNWGCKISMOH-UHFFFAOYSA-N |
Synonym | o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657 |
PubChem CID | 6940 |
IUPAC Name | 2-bromobenzoic acid |
SMILES | C1=CC=C(C(=C1)C(=O)O)Br |
Chemical Identifiers
88-65-3 | |
201.019 | |
XRXMNWGCKISMOH-UHFFFAOYSA-N | |
6940 | |
C1=CC=C(C(=C1)C(=O)O)Br |
C7H5BrO2 | |
MFCD00002402 | |
o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657 | |
2-bromobenzoic acid |
Specifications
88-65-3 | |
White-Yellow | |
MFCD00002402 | |
o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657 | |
C1=CC=C(C(=C1)C(=O)O)Br | |
201.019 | |
201.02 | |
Crystalline Powder |
150°C | |
C7H5BrO2 | |
25 g | |
XRXMNWGCKISMOH-UHFFFAOYSA-N | |
2-bromobenzoic acid | |
6940 | |
≥98.0% (GC,T) | |
2-Bromobenzoic Acid |
Safety and Handling
EINECSNumber : (3)-0062
RTECSNumber : DG4448035
TSCA : Yes