missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 106935000

 View more versions of this product

Catalog No. AC106935000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromoethylamine hydrobromide
2576-47-8,107-09-5
100.0
White to Beige
98.5% min. (Argentometry)
C2H6BrN·HBr
MFCD00012886
04,134
Solubility in water: >500g/L (20°C)
C(CBr)N.Br
204.9
204.9
Crystalline Powder or Crystals
99%
98.5
172.0°C to 176.0°C
Authentic
Plastic bottle
BrCH2CH2NH2·HBr
500 g
2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
WJAXXWSZNSFVNG-UHFFFAOYSA-N
2-bromoethanamine;hydrobromide
2774217
99%

Chemical Identifiers

2576-47-8
204.9
WJAXXWSZNSFVNG-UHFFFAOYSA-N
2774217
C(CBr)N.Br
C2H6BrN·HBr
MFCD00012886
2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
2-bromoethanamine;hydrobromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.<br

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 219-924-2

RTECSNumber : KQ8225000

TSCA : TSCA

Recommended Storage : this product has a strong avidity for water

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only