Learn More
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
$98.25 - $651.11
Chemical Identifiers
CAS | 2576-47-8 |
---|---|
Molecular Formula | C2H6BrN·HBr |
Molecular Weight (g/mol) | 204.9 |
MDL Number | MFCD00012886 |
InChI Key | WJAXXWSZNSFVNG-UHFFFAOYSA-N |
Synonym | 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide |
PubChem CID | 2774217 |
IUPAC Name | 2-bromoethanamine;hydrobromide |
SMILES | C(CBr)N.Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC106930250
|
Thermo Scientific Chemicals
106930250 |
25 g | Plastic bottle |
Each for $98.25
|
|
||||
AC106931000
|
Thermo Scientific Chemicals
106931000 |
100 g | Plastic bottle |
Each for $219.31
|
|
||||
AC106935000
|
Thermo Scientific Chemicals
106935000 |
500 g | Plastic bottle |
Each for $651.11
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2576-47-8 | |
204.9 | |
WJAXXWSZNSFVNG-UHFFFAOYSA-N | |
2774217 | |
C(CBr)N.Br |
C2H6BrN·HBr | |
MFCD00012886 | |
2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide | |
2-bromoethanamine;hydrobromide |
Specifications
2576-47-8,107-09-5 | |
100.0 | |
White to Beige | |
98.5% min. (Argentometry) | |
C2H6BrN·HBr | |
MFCD00012886 | |
04,134 | |
Solubility in water: >500g/L (20°C) | |
C(CBr)N.Br | |
204.9 | |
204.9 | |
Crystalline Powder or Crystals |
98.5 | |
172.0°C to 176.0°C | |
Authentic | |
Plastic bottle | |
BrCH2CH2NH2·HBr | |
25 g | |
2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide | |
WJAXXWSZNSFVNG-UHFFFAOYSA-N | |
2-bromoethanamine;hydrobromide | |
2774217 | |
99% | |
2-Bromoethylamine hydrobromide, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.<br
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 219-924-2
RTECSNumber : KQ8225000
TSCA : TSCA
Recommended Storage : this product has a strong avidity for water
RUO – Research Use Only