missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromomalonaldehyde, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 397900250

 View more versions of this product

Catalog No. AC397900250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromomalonaldehyde
132.0°C to 136.0°C
Glass bottle
BrCH(CHO)2
2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
C(=O)C(C=O)Br
150.95
150.95
2065-75-0
98.5 % min. (Acid)
C3H3BrO2
25 g
SURMYNZXHKLDFO-UHFFFAOYSA-N
2-bromopropanedial
74945
97+%

Chemical Identifiers

2065-75-0
150.95
2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
2-bromopropanedial
C3H3BrO2
SURMYNZXHKLDFO-UHFFFAOYSA-N
74945
C(=O)C(C=O)Br
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only