Learn More
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
$70.76 - $227.24
Chemical Identifiers
CAS | 31106-82-8 |
---|---|
Molecular Formula | C6H6BrN·HBr |
Molecular Weight (g/mol) | 252.94 |
MDL Number | MFCD01863544 |
InChI Key | JQDNCGRNPYKRAO-UHFFFAOYSA-N |
Synonym | 2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide |
PubChem CID | 2734720 |
IUPAC Name | 2-(bromomethyl)pyridine;hydrobromide |
SMILES | C1=CC=NC(=C1)CBr.Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC370110050
|
Thermo Scientific Chemicals
370110050 |
5 g | Glass bottle |
Each for $70.76
|
|
||||
AC370110250
|
Thermo Scientific Chemicals
370110250 |
25 g | Glass bottle |
Each for $227.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
31106-82-8 | |
252.94 | |
JQDNCGRNPYKRAO-UHFFFAOYSA-N | |
2734720 | |
C1=CC=NC(=C1)CBr.Br |
C6H6BrN·HBr | |
MFCD01863544 | |
2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide | |
2-(bromomethyl)pyridine;hydrobromide |
Specifications
31106-82-8 | |
100.0 | |
Brown to White | |
98% | |
C6H6BrN·HBr | |
MFCD01863544 | |
2-bromomethyl pyridine hydrobromide, 2-picolyl bromide hydrobromide, 2-bromomethyl pyridine hbr, pyridine, 2-bromomethyl-, hydrobromide, 2-bromomethyl pyridine, bromide, c6h6brn.hbr, acmc-20aim5, ksc495g6r, 2-bromomethylpyridine hydrobromide, bromomethyl pyridine hydrobromide | |
C1=CC=NC(=C1)CBr.Br | |
252.94 | |
252.94 | |
Crystalline Powder or Crystals |
97.5 | |
149.0°C to 152.0°C | |
Authentic | |
Glass bottle | |
BrCH2(C5H3N) | |
5 g | |
JQDNCGRNPYKRAO-UHFFFAOYSA-N | |
2-(bromomethyl)pyridine;hydrobromide | |
2734720 | |
98% | |
2-(Bromomethyl)pyridine hydrobromide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
RUO – Research Use Only