Learn More
2-Butenylmagnesium chloride, 0.5M in MeTHF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H51171AE
Description
2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications
2-Butenylmagnesium chloride | |
C4H7ClMg | |
MFCD01321226 | |
UN3399 | |
2-butenylmagnesium chloride, 2-butenylmagnesiumchloride,0.5minmethf, 2-butenylmagnesium chloride, 0.5m in methf, 2e-but-2-en-1-yl chloro magnesium, 2-butenylmagnesium chloride solution, 0.5 m in thf | |
XQAMFOKVRXJOGD-CZEFNJPISA-M | |
magnesium;(E)-but-2-ene;chloride | |
6104506 | |
0.5M in MeTHF |
6088-88-6 | |
CH3CH=CHCH2MgCl | |
100 mL | |
Air and moisture sensitive | |
It reacts violently with water. | |
CC=C[CH2-].[Mg+2].[Cl-] | |
114.855 | |
114.86 |
Chemical Identifiers
6088-88-6 | |
MFCD01321226 | |
6104506 |
C4H7ClMg | |
2-butenylmagnesium chloride, 2-butenylmagnesiumchloride,0.5minmethf, 2-butenylmagnesium chloride, 0.5m in methf, 2e-but-2-en-1-yl chloro magnesium, 2-butenylmagnesium chloride solution, 0.5 m in thf |
Safety and Handling
GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only