Learn More
2-Butenylmagnesium chloride, 0.5M in MeTHF, Thermo Scientific Chemicals
$477.05 - $477.05
Chemical Identifiers
CAS | 6088-88-6 |
---|---|
Molecular Formula | C4H7ClMg |
MDL Number | MFCD01321226 |
Synonym | 2-butenylmagnesium chloride, 2-butenylmagnesiumchloride,0.5minmethf, 2-butenylmagnesium chloride, 0.5m in methf, 2e-but-2-en-1-yl chloro magnesium, 2-butenylmagnesium chloride solution, 0.5 m in thf |
PubChem CID | 6104506 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH51171AE
|
Thermo Scientific Chemicals
H51171AE |
100 mL |
Each for $477.05
|
|
|||||
Description
2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Butenylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
Chemical Identifiers
6088-88-6 | |
MFCD01321226 | |
6104506 |
C4H7ClMg | |
2-butenylmagnesium chloride, 2-butenylmagnesiumchloride,0.5minmethf, 2-butenylmagnesium chloride, 0.5m in methf, 2e-but-2-en-1-yl chloro magnesium, 2-butenylmagnesium chloride solution, 0.5 m in thf |
Specifications
6088-88-6 | |
CH3CH=CHCH2MgCl | |
100 mL | |
Air and moisture sensitive | |
It reacts violently with water. | |
CC=C[CH2-].[Mg+2].[Cl-] | |
114.855 | |
114.86 | |
2-Butenylmagnesium chloride |
C4H7ClMg | |
MFCD01321226 | |
UN3399 | |
2-butenylmagnesium chloride, 2-butenylmagnesiumchloride,0.5minmethf, 2-butenylmagnesium chloride, 0.5m in methf, 2e-but-2-en-1-yl chloro magnesium, 2-butenylmagnesium chloride solution, 0.5 m in thf | |
XQAMFOKVRXJOGD-CZEFNJPISA-M | |
magnesium;(E)-but-2-ene;chloride | |
6104506 | |
0.5M in MeTHF |
Safety and Handling
GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only