missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butynoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 308060010

 View more versions of this product

Catalog No. AC308060010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Butynoic acid
97.5
74.0°C to 78.0°C
Authentic
Glass bottle
CH3C≡CCO2H
1 g
LUEHNHVFDCZTGL-UHFFFAOYSA-N
but-2-ynoic acid
68535
98%
590-93-2
100.0
White to Yellow
98%
C4H4O2
MFCD00004363
2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid
CC#CC(O)=O
84.07
84.07
Powder

Chemical Identifiers

590-93-2
84.07
LUEHNHVFDCZTGL-UHFFFAOYSA-N
68535
CC#CC(O)=O
C4H4O2
MFCD00004363
2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid
but-2-ynoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 209-695-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only