missing translation for 'onlineSavingsMsg'
Learn More

2-Butynoic acid, 98%

CAS: 590-93-2 | C4H4O2 | 84.07 g/mol

$87.45 - $87.45

Chemical Identifiers

CAS 590-93-2
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00004363
InChI Key LUEHNHVFDCZTGL-UHFFFAOYSA-N
Synonym 2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid
PubChem CID 68535
IUPAC Name but-2-ynoic acid
SMILES CC#CC(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC308060010
SDS View Documents
Thermo Scientific Chemicals
308060010
1 g Glass bottle
Each for $87.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

590-93-2
84.07
LUEHNHVFDCZTGL-UHFFFAOYSA-N
68535
CC#CC(O)=O
C4H4O2
MFCD00004363
2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid
but-2-ynoic acid

Specifications

590-93-2
100.0
White to Yellow
98%
C4H4O2
MFCD00004363
2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid
CC#CC(O)=O
84.07
84.07
Powder
97.5
74.0°C to 78.0°C
Authentic
Glass bottle
CH3C≡CCO2H
1 g
LUEHNHVFDCZTGL-UHFFFAOYSA-N
but-2-ynoic acid
68535
98%
2-Butynoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 209-695-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only