missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-1-iodo-4-nitrobenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1934014

 View more versions of this product

Catalog No. AAA1934014


Explore available promotions

Description

Description

3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications

Specifications

2-Chloro-1-iodo-4-nitrobenzene
97°C to 101°C
MFCD00024589
2502411
3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene
FQXFHRSYMORXKN-UHFFFAOYSA-N
2-chloro-1-iodo-4-nitrobenzene
96640
98%
41252-96-4
C6H3ClINO2
25 g
Light sensitive
Slightly soluble in water.
C1=CC(=C(C=C1[N+](=O)[O-])Cl)I
283.449
283.45

Chemical Identifiers

41252-96-4
283.449
FQXFHRSYMORXKN-UHFFFAOYSA-N
96640
C1=CC(=C(C=C1[N+](=O)[O-])Cl)I
C6H3ClINO2
MFCD00024589
3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene
2-chloro-1-iodo-4-nitrobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 255-278-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only