Learn More
2-Chloro-1-iodo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
$69.77 - $960.58
Chemical Identifiers
CAS | 41252-96-4 |
---|---|
Molecular Formula | C6H3ClINO2 |
Molecular Weight (g/mol) | 283.449 |
MDL Number | MFCD00024589 |
InChI Key | FQXFHRSYMORXKN-UHFFFAOYSA-N |
Synonym | 3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene |
PubChem CID | 96640 |
IUPAC Name | 2-chloro-1-iodo-4-nitrobenzene |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])Cl)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1934003
|
Thermo Scientific Chemicals
A1934003 |
1 g |
Each for $69.77
|
|
||||||
AAA1934006
|
Thermo Scientific Chemicals
A1934006 |
5 g |
Each for $231.92
|
|
||||||
AAA1934014
|
Thermo Scientific Chemicals
A1934014 |
25 g |
Each for $960.58
|
|
||||||
Description
3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-chloro-4-trifluoromethylanilinewas prepared in two steps by reaction of trifluoromethylcopper with 3-chloro-4-iodonitrobenzene.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Chemical Identifiers
41252-96-4 | |
283.449 | |
FQXFHRSYMORXKN-UHFFFAOYSA-N | |
96640 | |
C1=CC(=C(C=C1[N+](=O)[O-])Cl)I |
C6H3ClINO2 | |
MFCD00024589 | |
3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene | |
2-chloro-1-iodo-4-nitrobenzene |
Specifications
41252-96-4 | |
C6H3ClINO2 | |
1 g | |
Light sensitive | |
Slightly soluble in water. | |
C1=CC(=C(C=C1[N+](=O)[O-])Cl)I | |
283.449 | |
283.45 | |
2-Chloro-1-iodo-4-nitrobenzene |
97°C to 101°C | |
MFCD00024589 | |
2502411 | |
3-chloro-4-iodonitrobenzene, 3-chloro-4-iodo-nitrobenzene, 3-chloro-4-iodo-nitro-benzene, benzene, 2-chloro-1-iodo-4-nitro, 2-chlor-1-iod-4-nitrobenzol, acmc-20a36k, 3-chloro-4-iodonitro benzene, 1-iodo-2-chloro-4-nitrobenzene, 2-chloro-1-iodo-4-nitro-benzene, 1-chloro-2-iodo-5-nitrobenzene | |
FQXFHRSYMORXKN-UHFFFAOYSA-N | |
2-chloro-1-iodo-4-nitrobenzene | |
96640 | |
98% |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 255-278-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only