missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J60927MD

 View more versions of this product

Catalog No. AAJ60927MD


Explore available promotions

Description

Description

A purine nucleoside anti-metabolite. Toxic to nondividing lymphocytes and monocytes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A purine nucleoside anti-metabolite
  • Toxic to nondividing lymphocytes and monocytes
Specifications

Specifications

2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine
White
MFCD00871077
UN2811
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1
285.69
CHEBI:681569
≥99%
123318-82-1
C10H12ClN5O3
250 mg
14,2372
ADXUXRNLBYKGOA-GPGUAWMRNA-N
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
119182
303.68
Powder

Chemical Identifiers

123318-82-1
285.69
ADXUXRNLBYKGOA-GPGUAWMRNA-N
119182
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
C10H12ClN5O3
MFCD00871077
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
CHEBI:681569
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : UD7473000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only