Learn More
2-Chloro-3',4'-dihydroxyacetophenone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2349622
Description
2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.
Solubility
Soluble in DMSO, methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
2-Chloro-3',4'-dihydroxyacetophenone | |
172°C to 176°C | |
MFCD00002200 | |
2092660 | |
Soluble in DMSO,methanol. | |
C1=CC(=C(C=C1C(=O)CCl)O)O | |
186.591 | |
CHEBI:51844 | |
97% |
99-40-1 | |
C8H7ClO3 | |
100 g | |
2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone | |
LWTJEJCZJFZKEL-UHFFFAOYSA-N | |
2-chloro-1-(3,4-dihydroxyphenyl)ethanone | |
66834 | |
186.59 |
Chemical Identifiers
99-40-1 | |
186.591 | |
LWTJEJCZJFZKEL-UHFFFAOYSA-N | |
66834 | |
2-chloro-1-(3,4-dihydroxyphenyl)ethanone |
C8H7ClO3 | |
MFCD00002200 | |
2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone | |
CHEBI:51844 | |
C1=CC(=C(C=C1C(=O)CCl)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-754-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only