missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-3',4'-dihydroxyacetophenone, 97%, Thermo Scientific Chemicals

$101.52 - $958.74

Chemical Identifiers

CAS 99-40-1
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00002200
InChI Key LWTJEJCZJFZKEL-UHFFFAOYSA-N
Synonym 2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone
PubChem CID 66834
ChEBI CHEBI:51844
IUPAC Name 2-chloro-1-(3,4-dihydroxyphenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CCl)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2349614
View Documents
Thermo Scientific Chemicals
B2349614
25 g
Each for $101.52
 
AAB2349622
View Documents
Thermo Scientific Chemicals
B2349622
100 g
Each for $294.01
 
AAB2349636
View Documents
Thermo Scientific Chemicals
B2349636
500 g
Each for $958.74
 
Description

Description

2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

Solubility
Soluble in DMSO, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

99-40-1
186.591
LWTJEJCZJFZKEL-UHFFFAOYSA-N
66834
2-chloro-1-(3,4-dihydroxyphenyl)ethanone
C8H7ClO3
MFCD00002200
2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone
CHEBI:51844
C1=CC(=C(C=C1C(=O)CCl)O)O

Specifications

99-40-1
C8H7ClO3
25 g
2-chloro-3',4'-dihydroxyacetophenone, 2-chloro-1-3,4-dihydroxyphenyl ethanone, 4-chloroacetyl catechol, 3,4-dihydroxyphenacyl chloride, ethanone, 2-chloro-1-3,4-dihydroxyphenyl, unii-fn6949l56q, 3',4'-dihydroxy-2-chloroacetophenone, chloromethyl 3,4-dihydroxyphenyl ketone, 2-chloro-3,4-dihydroxyacetophenone, .alpha.-chloro-3,4-dihydroxyacetophenone
LWTJEJCZJFZKEL-UHFFFAOYSA-N
2-chloro-1-(3,4-dihydroxyphenyl)ethanone
66834
186.59
2-Chloro-3',4'-dihydroxyacetophenone
172°C to 176°C
MFCD00002200
2092660
Soluble in DMSO,methanol.
C1=CC(=C(C=C1C(=O)CCl)O)O
186.591
CHEBI:51844
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-754-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only