missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-3-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 241790050
Specifications
2-chloro-3-(trifluoromethyl)pyridine | |
96.0 | |
166.0°C to 168.0°C | |
Authentic | |
Glass bottle | |
5g | |
Solubility in water: insoluble. Other solubilities: reacts at high temperature with strong acids and,alkalis | |
C1=CC(=C(N=C1)Cl)C(F)(F)F | |
181.54 | |
181.54 |
65753-47-1 | |
100.0 | |
82°C | |
96% min. (GC) | |
C6H3ClF3N | |
2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t | |
RXATZPCCMYMPME-UHFFFAOYSA-N | |
2-chloro-3-(trifluoromethyl)pyridine | |
589833 | |
97% |
Chemical Identifiers
65753-47-1 | |
181.54 | |
2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t | |
2-chloro-3-(trifluoromethyl)pyridine |
C6H3ClF3N | |
RXATZPCCMYMPME-UHFFFAOYSA-N | |
589833 | |
C1=CC(=C(N=C1)Cl)C(F)(F)F |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Harmful to aquatic life with long lasting e
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
RUO â Research Use Only