missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-3-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 65753-47-1 |
---|---|
Molecular Formula | C6H3ClF3N |
Molecular Weight (g/mol) | 181.54 |
InChI Key | RXATZPCCMYMPME-UHFFFAOYSA-N |
Synonym | 2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t |
PubChem CID | 589833 |
IUPAC Name | 2-chloro-3-(trifluoromethyl)pyridine |
SMILES | C1=CC(=C(N=C1)Cl)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC241790050
|
Thermo Scientific Chemicals
241790050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC241790250
|
Thermo Scientific Chemicals
241790250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
65753-47-1 | |
181.54 | |
2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t | |
2-chloro-3-(trifluoromethyl)pyridine |
C6H3ClF3N | |
RXATZPCCMYMPME-UHFFFAOYSA-N | |
589833 | |
C1=CC(=C(N=C1)Cl)C(F)(F)F |
Specifications
65753-47-1 | |
100.0 | |
82°C | |
96% min. (GC) | |
C6H3ClF3N | |
2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t | |
RXATZPCCMYMPME-UHFFFAOYSA-N | |
2-chloro-3-(trifluoromethyl)pyridine | |
589833 | |
97% |
96.0 | |
166.0°C to 168.0°C | |
Authentic | |
Glass bottle | |
5g | |
Solubility in water: insoluble. Other solubilities: reacts at high temperature with strong acids and,alkalis | |
C1=CC(=C(N=C1)Cl)C(F)(F)F | |
181.54 | |
181.54 | |
2-chloro-3-(trifluoromethyl)pyridine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Harmful to aquatic life with long lasting e
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
RUO â Research Use Only