missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2759114

 View more versions of this product

Catalog No. AAH2759114


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-4-fluorobenzeneboronic acid
C6H5BClFO2
25 g
XOFNMNLYGPKKOV-UHFFFAOYSA-N
(2-chloro-4-fluorophenyl)boronic acid
17750229
98%
313545-72-1
MFCD02684295
2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
OB(O)C1=CC=C(F)C=C1Cl
174.36
174.37

Chemical Identifiers

313545-72-1
174.36
XOFNMNLYGPKKOV-UHFFFAOYSA-N
17750229
OB(O)C1=CC=C(F)C=C1Cl
C6H5BClFO2
MFCD02684295
2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
(2-chloro-4-fluorophenyl)boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only