missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

$171.66 - $1241.79

Chemical Identifiers

CAS 313545-72-1
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD02684295
InChI Key XOFNMNLYGPKKOV-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
PubChem CID 17750229
IUPAC Name (2-chloro-4-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=C(F)C=C1Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2759103
View Documents
Thermo Scientific Chemicals
H2759103
1 g
Each for $171.66
 
AAH2759106
View Documents
Thermo Scientific Chemicals
H2759106
5 g
Each for $445.59
 
AAH2759114
View Documents
Thermo Scientific Chemicals
H2759114
25 g
Each for $1,241.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

313545-72-1
174.36
XOFNMNLYGPKKOV-UHFFFAOYSA-N
17750229
OB(O)C1=CC=C(F)C=C1Cl
C6H5BClFO2
MFCD02684295
2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
(2-chloro-4-fluorophenyl)boronic acid

Specifications

313545-72-1
MFCD02684295
2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
OB(O)C1=CC=C(F)C=C1Cl
174.36
174.37
2-Chloro-4-fluorobenzeneboronic acid
C6H5BClFO2
1 g
XOFNMNLYGPKKOV-UHFFFAOYSA-N
(2-chloro-4-fluorophenyl)boronic acid
17750229
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only