missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-fluorobenzylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1295706

 View more versions of this product

Catalog No. AAL1295706


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-4-fluorobenzylamine
94°C to 95°C (15 mmHg)
C7H7ClFN
MFCD00042532
UN2735
Air Sensitive
CBKWAXKMZUULLO-UHFFFAOYSA-N
(2-chloro-4-fluorophenyl)methanamine
139909
97%
15205-11-5
>110°C (230°F)
1.535
5 g
2936421
2-chloro-4-fluorobenzylamine, 2-chloro-4-fluorophenyl methanamine, 2-chloro-4-fluorobenzyl amine, 1-2-chloro-4-fluorophenyl methanamine, benzenemethanamine, 2-chloro-4-fluoro, 2-chloro-4-fluorophenyl methylamine, 2-chloro-4-florobenzylamine, acmc-1c6hl, 2-chloro4-fluorobenzylamine, 2-chloro-4-fluorobenzylamide
C1=CC(=C(C=C1F)Cl)CN
159.588
159.59

Chemical Identifiers

15205-11-5
159.588
CBKWAXKMZUULLO-UHFFFAOYSA-N
139909
C1=CC(=C(C=C1F)Cl)CN
C7H7ClFN
MFCD00042532
2-chloro-4-fluorobenzylamine, 2-chloro-4-fluorophenyl methanamine, 2-chloro-4-fluorobenzyl amine, 1-2-chloro-4-fluorophenyl methanamine, benzenemethanamine, 2-chloro-4-fluoro, 2-chloro-4-fluorophenyl methylamine, 2-chloro-4-florobenzylamine, acmc-1c6hl, 2-chloro4-fluorobenzylamine, 2-chloro-4-fluorobenzylamide
(2-chloro-4-fluorophenyl)methanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only