Learn More
2-Chloro-4-fluorobenzylamine, 97%, Thermo Scientific Chemicals
$152.25 - $506.26
Chemical Identifiers
CAS | 15205-11-5 |
---|---|
Molecular Formula | C7H7ClFN |
Molecular Weight (g/mol) | 159.588 |
MDL Number | MFCD00042532 |
InChI Key | CBKWAXKMZUULLO-UHFFFAOYSA-N |
Synonym | 2-chloro-4-fluorobenzylamine, 2-chloro-4-fluorophenyl methanamine, 2-chloro-4-fluorobenzyl amine, 1-2-chloro-4-fluorophenyl methanamine, benzenemethanamine, 2-chloro-4-fluoro, 2-chloro-4-fluorophenyl methylamine, 2-chloro-4-florobenzylamine, acmc-1c6hl, 2-chloro4-fluorobenzylamine, 2-chloro-4-fluorobenzylamide |
PubChem CID | 139909 |
IUPAC Name | (2-chloro-4-fluorophenyl)methanamine |
SMILES | C1=CC(=C(C=C1F)Cl)CN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1295703
|
Thermo Scientific Chemicals
L1295703 |
1 g |
Each for $152.25
|
|
|||||
AAL1295706
|
Thermo Scientific Chemicals
L1295706 |
5 g |
Each for $506.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15205-11-5 | |
159.588 | |
CBKWAXKMZUULLO-UHFFFAOYSA-N | |
139909 | |
C1=CC(=C(C=C1F)Cl)CN |
C7H7ClFN | |
MFCD00042532 | |
2-chloro-4-fluorobenzylamine, 2-chloro-4-fluorophenyl methanamine, 2-chloro-4-fluorobenzyl amine, 1-2-chloro-4-fluorophenyl methanamine, benzenemethanamine, 2-chloro-4-fluoro, 2-chloro-4-fluorophenyl methylamine, 2-chloro-4-florobenzylamine, acmc-1c6hl, 2-chloro4-fluorobenzylamine, 2-chloro-4-fluorobenzylamide | |
(2-chloro-4-fluorophenyl)methanamine |
Specifications
15205-11-5 | |
>110°C (230°F) | |
1.535 | |
1 g | |
2936421 | |
2-chloro-4-fluorobenzylamine, 2-chloro-4-fluorophenyl methanamine, 2-chloro-4-fluorobenzyl amine, 1-2-chloro-4-fluorophenyl methanamine, benzenemethanamine, 2-chloro-4-fluoro, 2-chloro-4-fluorophenyl methylamine, 2-chloro-4-florobenzylamine, acmc-1c6hl, 2-chloro4-fluorobenzylamine, 2-chloro-4-fluorobenzylamide | |
C1=CC(=C(C=C1F)Cl)CN | |
159.588 | |
159.59 | |
2-Chloro-4-fluorobenzylamine |
94°C to 95°C (15 mmHg) | |
C7H7ClFN | |
MFCD00042532 | |
UN2735 | |
Air Sensitive | |
CBKWAXKMZUULLO-UHFFFAOYSA-N | |
(2-chloro-4-fluorophenyl)methanamine | |
139909 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only