Learn More
2-Chloro-4-nitrophenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2156130
Description
2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.
Solubility
Slightly Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
Specifications
2-Chloro-4-nitrophenol | |
108°C to 112°C | |
C6H4ClNO3 | |
250 g | |
2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908 | |
BOFRXDMCQRTGII-UHFFFAOYSA-N | |
173.552 | |
173.56 |
619-08-9 | |
Characteristic | |
MFCD00043910 | |
2046372 | |
Slightly Soluble in water. | |
C1=CC(=C(C=C1[N+](=O)[O-])Cl)O | |
12074 | |
97% |
Chemical Identifiers
619-08-9 | |
173.552 | |
BOFRXDMCQRTGII-UHFFFAOYSA-N | |
12074 |
C6H4ClNO3 | |
MFCD00043910 | |
2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908 | |
C1=CC(=C(C=C1[N+](=O)[O-])Cl)O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 210-578-8
RTECSNumber : SK5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only