missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-4-nitrophenol, 97%, Thermo Scientific Chemicals

$66.66 - $662.90

Chemical Identifiers

CAS 619-08-9
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.552
MDL Number MFCD00043910
InChI Key BOFRXDMCQRTGII-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908
PubChem CID 12074
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2156109
View Documents
Thermo Scientific Chemicals
B2156109
10 g N/A
 
AAB2156118
View Documents
Thermo Scientific Chemicals
B2156118
50 g N/A
 
AAB2156130
View Documents
Thermo Scientific Chemicals
B2156130
250 g N/A
 
Description

Description

2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.

Solubility
Slightly Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
Specifications

Chemical Identifiers

619-08-9
173.552
BOFRXDMCQRTGII-UHFFFAOYSA-N
12074
C6H4ClNO3
MFCD00043910
2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908
C1=CC(=C(C=C1[N+](=O)[O-])Cl)O

Specifications

619-08-9
Characteristic
MFCD00043910
2046372
Slightly Soluble in water.
C1=CC(=C(C=C1[N+](=O)[O-])Cl)O
12074
97%
108°C to 112°C
C6H4ClNO3
10 g
2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908
BOFRXDMCQRTGII-UHFFFAOYSA-N
173.552
173.56
2-Chloro-4-nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 210-578-8

RTECSNumber : SK5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only