missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-4-(trifluoromethyl)anisole, 97%, Thermo Scientific™
$335.41 - $335.41
Chemical Identifiers
CAS | 1214340-18-7 |
---|---|
Molecular Formula | C8H6ClF3O |
Molecular Weight (g/mol) | 210.58 |
MDL Number | MFCD13185462 |
InChI Key | VVRXOIBFPBBYRU-UHFFFAOYSA-N |
Synonym | 3-chloro-4-methoxybenzotrifluoride, 2-chloro-1-methoxy-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl anisole, 2-methoxy-5-trifluoromethyl chlorobenzene, 2-chloro-4-trifluoromethyl phenyl methyl ether |
PubChem CID | 54413375 |
IUPAC Name | 2-chloro-1-methoxy-4-(trifluoromethyl)benzene |
SMILES | COC1=C(Cl)C=C(C=C1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3355403
|
Thermo Scientific Chemicals
H3355403 |
1 g |
Each for $335.41
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Chemical Identifiers
1214340-18-7 | |
210.58 | |
VVRXOIBFPBBYRU-UHFFFAOYSA-N | |
54413375 | |
COC1=C(Cl)C=C(C=C1)C(F)(F)F |
C8H6ClF3O | |
MFCD13185462 | |
3-chloro-4-methoxybenzotrifluoride, 2-chloro-1-methoxy-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl anisole, 2-methoxy-5-trifluoromethyl chlorobenzene, 2-chloro-4-trifluoromethyl phenyl methyl ether | |
2-chloro-1-methoxy-4-(trifluoromethyl)benzene |
Specifications
1214340-18-7 | |
1.4755 | |
1 g | |
Sparingly soluble in water.(0.26g/L) (25°C), | |
COC1=C(Cl)C=C(C=C1)C(F)(F)F | |
210.58 | |
210.58 | |
2-Chloro-4-(trifluoromethyl)anisole |
C8H6ClF3O | |
MFCD13185462 | |
3-chloro-4-methoxybenzotrifluoride, 2-chloro-1-methoxy-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethyl anisole, 2-methoxy-5-trifluoromethyl chlorobenzene, 2-chloro-4-trifluoromethyl phenyl methyl ether | |
VVRXOIBFPBBYRU-UHFFFAOYSA-N | |
2-chloro-1-methoxy-4-(trifluoromethyl)benzene | |
54413375 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures