missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-ethylpyrimidine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2685606

 View more versions of this product

Catalog No. AAH2685606


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-5-ethylpyrimidine
1.174
>110°C (230°F)
1.521
5 g
5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
CCC1=CN=C(Cl)N=C1
142.59
142.59
111196-81-7
223°C
C6H7ClN2
MFCD00799503
Moisture sensitive; May form a precipitate
BGLLZQRUXJGTAD-UHFFFAOYSA-N
2-chloro-5-ethylpyrimidine
3572763
98%

Chemical Identifiers

111196-81-7
142.59
BGLLZQRUXJGTAD-UHFFFAOYSA-N
3572763
CCC1=CN=C(Cl)N=C1
C6H7ClN2
MFCD00799503
5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
2-chloro-5-ethylpyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only