missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-5-ethylpyrimidine, 98%, Thermo Scientific Chemicals

$157.10 - $439.08

Chemical Identifiers

CAS 111196-81-7
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00799503
InChI Key BGLLZQRUXJGTAD-UHFFFAOYSA-N
Synonym 5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
PubChem CID 3572763
IUPAC Name 2-chloro-5-ethylpyrimidine
SMILES CCC1=CN=C(Cl)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2685606
View Documents
Thermo Scientific Chemicals
H2685606
5 g
Each for $157.10
 
AAH2685614
View Documents
Thermo Scientific Chemicals
H2685614
25 g
Each for $439.08
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111196-81-7
142.59
BGLLZQRUXJGTAD-UHFFFAOYSA-N
3572763
CCC1=CN=C(Cl)N=C1
C6H7ClN2
MFCD00799503
5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
2-chloro-5-ethylpyrimidine

Specifications

111196-81-7
223°C
C6H7ClN2
MFCD00799503
Moisture sensitive; May form a precipitate
BGLLZQRUXJGTAD-UHFFFAOYSA-N
2-chloro-5-ethylpyrimidine
3572763
98%
1.174
>110°C (230°F)
1.521
5 g
5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
CCC1=CN=C(Cl)N=C1
142.59
142.59
2-Chloro-5-ethylpyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only