missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Chloro-5'-(trifluoromethyl)acetophenone, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2352406

 View more versions of this product

Catalog No. AAB2352406

Explore available promotions

Description

Description

2'-Chloro-5'-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Chloro-5′-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.11g/L @ 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications

Specifications

2'-Chloro-5'-(trifluoromethyl)acetophenone
C9H6ClF3O
5 g
Sparingly soluble in water 0.11g/L @ 25°C.
CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl
222.591
222.6
71648-45-8
MFCD00045092
2'-chloro-5'-trifluoromethyl acetophenone, 1-2-chloro-5-trifluoromethyl phenyl ethanone, 1-2-chloro-5-trifluoromethyl phenyl ethan-1-one, 2-chloro-5-trifluoromethyl acetophenone, acetophenone, 2-chloro-5-trifluoromethyl, 3-acetyl-4-chlorobenzotrifluoride, ethanone, 1-2-chloro-5-trifluoromethyl phenyl, 1-acetyl-2-chloro-5-trifluoromethyl benzene, pubchem4210
YRGBMTWHOFQSDJ-UHFFFAOYSA-N
1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone
144502
≥97%

Chemical Identifiers

71648-45-8
222.591
YRGBMTWHOFQSDJ-UHFFFAOYSA-N
144502
CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl
C9H6ClF3O
MFCD00045092
2'-chloro-5'-trifluoromethyl acetophenone, 1-2-chloro-5-trifluoromethyl phenyl ethanone, 1-2-chloro-5-trifluoromethyl phenyl ethan-1-one, 2-chloro-5-trifluoromethyl acetophenone, acetophenone, 2-chloro-5-trifluoromethyl, 3-acetyl-4-chlorobenzotrifluoride, ethanone, 1-2-chloro-5-trifluoromethyl phenyl, 1-acetyl-2-chloro-5-trifluoromethyl benzene, pubchem4210
1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only