Learn More
2'-Chloro-5'-(trifluoromethyl)acetophenone, 96%, Thermo Scientific Chemicals
$93.29 - $313.71
Chemical Identifiers
CAS | 71648-45-8 |
---|---|
Molecular Formula | C9H6ClF3O |
Molecular Weight (g/mol) | 222.591 |
MDL Number | MFCD00045092 |
InChI Key | YRGBMTWHOFQSDJ-UHFFFAOYSA-N |
Synonym | 2'-chloro-5'-trifluoromethyl acetophenone, 1-2-chloro-5-trifluoromethyl phenyl ethanone, 1-2-chloro-5-trifluoromethyl phenyl ethan-1-one, 2-chloro-5-trifluoromethyl acetophenone, acetophenone, 2-chloro-5-trifluoromethyl, 3-acetyl-4-chlorobenzotrifluoride, ethanone, 1-2-chloro-5-trifluoromethyl phenyl, 1-acetyl-2-chloro-5-trifluoromethyl benzene, pubchem4210 |
PubChem CID | 144502 |
IUPAC Name | 1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone |
SMILES | CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2352403
|
Thermo Scientific Chemicals
B2352403 |
1 g |
N/A
|
|
|||||
AAB2352406
|
Thermo Scientific Chemicals
B2352406 |
5 g |
N/A
|
|
|||||
Description
2'-Chloro-5'-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Chloro-5′-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.
Solubility
Sparingly soluble in water 0.11g/L @ 25°C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Chemical Identifiers
71648-45-8 | |
222.591 | |
YRGBMTWHOFQSDJ-UHFFFAOYSA-N | |
144502 | |
CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl |
C9H6ClF3O | |
MFCD00045092 | |
2'-chloro-5'-trifluoromethyl acetophenone, 1-2-chloro-5-trifluoromethyl phenyl ethanone, 1-2-chloro-5-trifluoromethyl phenyl ethan-1-one, 2-chloro-5-trifluoromethyl acetophenone, acetophenone, 2-chloro-5-trifluoromethyl, 3-acetyl-4-chlorobenzotrifluoride, ethanone, 1-2-chloro-5-trifluoromethyl phenyl, 1-acetyl-2-chloro-5-trifluoromethyl benzene, pubchem4210 | |
1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone |
Specifications
71648-45-8 | |
MFCD00045092 | |
2'-chloro-5'-trifluoromethyl acetophenone, 1-2-chloro-5-trifluoromethyl phenyl ethanone, 1-2-chloro-5-trifluoromethyl phenyl ethan-1-one, 2-chloro-5-trifluoromethyl acetophenone, acetophenone, 2-chloro-5-trifluoromethyl, 3-acetyl-4-chlorobenzotrifluoride, ethanone, 1-2-chloro-5-trifluoromethyl phenyl, 1-acetyl-2-chloro-5-trifluoromethyl benzene, pubchem4210 | |
YRGBMTWHOFQSDJ-UHFFFAOYSA-N | |
1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone | |
144502 | |
≥97% |
C9H6ClF3O | |
1 g | |
Sparingly soluble in water 0.11g/L @ 25°C. | |
CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl | |
222.591 | |
222.6 | |
2'-Chloro-5'-(trifluoromethyl)acetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only