missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-fluorocinnamic acid, predominantly trans, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals L0746803

 View more versions of this product

Catalog No. AAL0746803


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Chloro-6-fluorocinnamic acid
392-22-3
C9H6ClFO2
1 g
2-chloro-6-fluorocinnamic acid, 3-2-chloro-6-fluorophenyl acrylic acid, 2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid, 2e-3-2-chloro-6-fluorophenyl acrylic acid, e-3-2-chloro-6-fluoro-phenyl-acrylic acid, 2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e, 3-2-chloro-6-fluorophenyl prop-2-enoic acid, pubchem3430, rarechem bk hw 0069
C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F
200.593
200.6
predominantly trans
164°C to 172°C
MFCD00051582
3129946
NDWALECYVLNBQG-SNAWJCMRSA-N
(E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid
5324668
98%

Chemical Identifiers

392-22-3
200.593
NDWALECYVLNBQG-SNAWJCMRSA-N
5324668
C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F
C9H6ClFO2
MFCD00051582
2-chloro-6-fluorocinnamic acid, 3-2-chloro-6-fluorophenyl acrylic acid, 2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid, 2e-3-2-chloro-6-fluorophenyl acrylic acid, e-3-2-chloro-6-fluoro-phenyl-acrylic acid, 2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e, 3-2-chloro-6-fluorophenyl prop-2-enoic acid, pubchem3430, rarechem bk hw 0069
(E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions