missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-6-fluorocinnamic acid, predominantly trans, 98%, Thermo Scientific™
$36.00 - $79.70
Chemical Identifiers
CAS | 392-22-3 |
---|---|
Molecular Formula | C9H6ClFO2 |
Molecular Weight (g/mol) | 200.593 |
MDL Number | MFCD00051582 |
InChI Key | NDWALECYVLNBQG-SNAWJCMRSA-N |
Synonym | 2-chloro-6-fluorocinnamic acid, 3-2-chloro-6-fluorophenyl acrylic acid, 2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid, 2e-3-2-chloro-6-fluorophenyl acrylic acid, e-3-2-chloro-6-fluoro-phenyl-acrylic acid, 2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e, 3-2-chloro-6-fluorophenyl prop-2-enoic acid, pubchem3430, rarechem bk hw 0069 |
PubChem CID | 5324668 |
IUPAC Name | (E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid |
SMILES | C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0746803
|
Thermo Scientific Chemicals
L0746803 |
1 g |
Each for $36.00
|
|
|||||
AAL0746806
|
Thermo Scientific Chemicals
L0746806 |
5 g |
Each for $79.70
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
392-22-3 | |
200.593 | |
NDWALECYVLNBQG-SNAWJCMRSA-N | |
5324668 | |
C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F |
C9H6ClFO2 | |
MFCD00051582 | |
2-chloro-6-fluorocinnamic acid, 3-2-chloro-6-fluorophenyl acrylic acid, 2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid, 2e-3-2-chloro-6-fluorophenyl acrylic acid, e-3-2-chloro-6-fluoro-phenyl-acrylic acid, 2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e, 3-2-chloro-6-fluorophenyl prop-2-enoic acid, pubchem3430, rarechem bk hw 0069 | |
(E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid |
Specifications
392-22-3 | |
C9H6ClFO2 | |
1 g | |
2-chloro-6-fluorocinnamic acid, 3-2-chloro-6-fluorophenyl acrylic acid, 2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid, 2e-3-2-chloro-6-fluorophenyl acrylic acid, e-3-2-chloro-6-fluoro-phenyl-acrylic acid, 2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e, 3-2-chloro-6-fluorophenyl prop-2-enoic acid, pubchem3430, rarechem bk hw 0069 | |
C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F | |
200.593 | |
200.6 | |
2-Chloro-6-fluorocinnamic acid, predominantly trans |
164°C to 172°C | |
MFCD00051582 | |
3129946 | |
NDWALECYVLNBQG-SNAWJCMRSA-N | |
(E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid | |
5324668 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures