missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloro-6-fluorocinnamic acid, predominantly trans, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0746806
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Chloro-6-fluorocinnamic acid | |
392-22-3 | |
C9H6ClFO2 | |
5 g | |
2-chloro-6-fluorocinnamic acid, 3-2-chloro-6-fluorophenyl acrylic acid, 2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid, 2e-3-2-chloro-6-fluorophenyl acrylic acid, e-3-2-chloro-6-fluoro-phenyl-acrylic acid, 2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e, 3-2-chloro-6-fluorophenyl prop-2-enoic acid, pubchem3430, rarechem bk hw 0069 | |
C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F | |
200.593 | |
200.6 |
predominantly trans | |
164°C to 172°C | |
MFCD00051582 | |
3129946 | |
NDWALECYVLNBQG-SNAWJCMRSA-N | |
(E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid | |
5324668 | |
98% |
Chemical Identifiers
392-22-3 | |
200.593 | |
NDWALECYVLNBQG-SNAWJCMRSA-N | |
5324668 | |
C1=CC(=C(C(=C1)Cl)C=CC(=O)O)F |
C9H6ClFO2 | |
MFCD00051582 | |
2-chloro-6-fluorocinnamic acid, 3-2-chloro-6-fluorophenyl acrylic acid, 2e-3-2-chloro-6-fluorophenyl prop-2-enoic acid, 2e-3-2-chloro-6-fluorophenyl acrylic acid, e-3-2-chloro-6-fluoro-phenyl-acrylic acid, 2-propenoic acid, 3-2-chloro-6-fluorophenyl-, 2e, 3-2-chloro-6-fluorophenyl prop-2-enoic acid, pubchem3430, rarechem bk hw 0069 | |
(E)-3-(2-chloro-6-fluorophenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures