missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-6-fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1046014

 View more versions of this product

Catalog No. AAA1046014


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-6-fluorophenylacetic acid
120°C to 124°C
MFCD00004319
4401142
GUAIAAXDEJZRBP-UHFFFAOYSA-N
2-(2-chloro-6-fluorophenyl)acetic acid
123464
98%
37777-76-7
C8H6ClFO2
25 g
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114
C1=CC(=C(C(=C1)Cl)CC(=O)O)F
188.582
188.59

Chemical Identifiers

37777-76-7
188.582
GUAIAAXDEJZRBP-UHFFFAOYSA-N
123464
C1=CC(=C(C(=C1)Cl)CC(=O)O)F
C8H6ClFO2
MFCD00004319
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114
2-(2-chloro-6-fluorophenyl)acetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 253-661-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only