missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 135440100

 View more versions of this product

Catalog No. AC135440100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-N-(2, 6-dimethylphenyl)acetamide
97.5
143.0°C to 148.0°C
Authentic
Glass bottle
(CH3)2C6H3NHC(=O)CH2Cl
12,III,2464
Solubility in water: insoluble
CC1=C(C(=CC=C1)C)NC(=O)CCl
197.66
197.66
Crystalline Powder or Needles
1131-01-7
100.0
White to Brown
97.5% min. (HPLC)
C10H12ClNO
10 g
2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx
FPQQSNUTBWFFLB-UHFFFAOYSA-N
2-chloro-N-(2,6-dimethylphenyl)acetamide
70798
98%

Chemical Identifiers

1131-01-7
197.66
2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx
2-chloro-N-(2,6-dimethylphenyl)acetamide
C10H12ClNO
FPQQSNUTBWFFLB-UHFFFAOYSA-N
70798
CC1=C(C(=CC=C1)C)NC(=O)CCl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face prot

GHS Signal Word: Warning

EINECSNumber : 214-460-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only