Learn More
2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%, Thermo Scientific Chemicals
$120.48 - $403.92
Chemical Identifiers
CAS | 1131-01-7 |
---|---|
Molecular Formula | C10H12ClNO |
Molecular Weight (g/mol) | 197.66 |
InChI Key | FPQQSNUTBWFFLB-UHFFFAOYSA-N |
Synonym | 2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx |
PubChem CID | 70798 |
IUPAC Name | 2-chloro-N-(2,6-dimethylphenyl)acetamide |
SMILES | CC1=C(C(=CC=C1)C)NC(=O)CCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC135440100
|
Thermo Scientific Chemicals
135440100 |
10 g | Glass bottle |
N/A
|
|
||||
AC135440500
|
Thermo Scientific Chemicals
135440500 |
50 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1131-01-7 | |
197.66 | |
2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx | |
2-chloro-N-(2,6-dimethylphenyl)acetamide |
C10H12ClNO | |
FPQQSNUTBWFFLB-UHFFFAOYSA-N | |
70798 | |
CC1=C(C(=CC=C1)C)NC(=O)CCl |
Specifications
1131-01-7 | |
100.0 | |
White to Brown | |
97.5% min. (HPLC) | |
C10H12ClNO | |
10 g | |
2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx | |
FPQQSNUTBWFFLB-UHFFFAOYSA-N | |
2-chloro-N-(2,6-dimethylphenyl)acetamide | |
70798 | |
98% | |
2-Chloro-N-(2, 6-dimethylphenyl)acetamide |
97.5 | |
143.0°C to 148.0°C | |
Authentic | |
Glass bottle | |
(CH3)2C6H3NHC(=O)CH2Cl | |
12,III,2464 | |
Solubility in water: insoluble | |
CC1=C(C(=CC=C1)C)NC(=O)CCl | |
197.66 | |
197.66 | |
Crystalline Powder or Needles |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face prot
GHS Signal Word: Warning
EINECSNumber : 214-460-7
RUO – Research Use Only