missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorobutane, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B21586AK

 View more versions of this product

Catalog No. AAB21586AK


Explore available promotions

Description

Description

2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

Solubility
Immiscible with water.

Notes
Incompatible with metals, copper, strong oxidizing agents and strong bases.
Specifications

Specifications

2-Chlorobutane
°C to 141 to°C to 140°C
67°C to 69°C
98+%
1.397
250 mL
1718770
sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl
BSPCSKHALVHRSR-UHFFFAOYSA-N
2-chlorobutane
6563
≥98%
78-86-4
0.873
-15°C (5°F)
C4H9Cl
MFCD00000871
UN1127
14,1561
Immiscible with water.
CCC(C)Cl
92.566
92.57
Liquid

Chemical Identifiers

78-86-4
92.566
BSPCSKHALVHRSR-UHFFFAOYSA-N
6563
CCC(C)Cl
C4H9Cl
MFCD00000871
sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl
2-chlorobutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P234-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P390-P501c

H225-H290

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES

EINECSNumber : 201-151-7

RTECSNumber : EJ6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only