missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorobutyric acid, 97%, Thermo Scientific Chemicals

$147.96 - $147.96

Chemical Identifiers

CAS 4170-24-5
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00021697
InChI Key RVBUZBPJAGZHSQ-UHFFFAOYSA-N
Synonym 2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro
PubChem CID 20137
IUPAC Name 2-chlorobutanoic acid
SMILES CCC(C(=O)O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2008314
View Documents
Thermo Scientific Chemicals
L2008314
25 g
Each for $147.96
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4170-24-5
122.548
RVBUZBPJAGZHSQ-UHFFFAOYSA-N
20137
CCC(C(=O)O)Cl
C4H7ClO2
MFCD00021697
2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro
2-chlorobutanoic acid

Specifications

4170-24-5
90°C to 92°C (12 mmHg)
C4H7ClO2
MFCD00021697
UN3265
2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro
CCC(C(=O)O)Cl
122.548
122.55
2-Chlorobutyric acid
1.19
112°C (233°F)
1.439
25 g
1720944
RVBUZBPJAGZHSQ-UHFFFAOYSA-N
2-chlorobutanoic acid
20137
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 224-029-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only