missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 404610050

 View more versions of this product

Catalog No. AC404610050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloroethanesulfonyl chloride
1622-32-8
100.0
1.5500g/mL
99°C
94% min. (ex Cl) (Argentometry)
C2H4Cl2O2S
ClCH2CH2SO2Cl
5 g
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
C(CCl)S(=O)(=O)Cl
163.02
163.02
Liquid
95%
94.0
Yellow to Brown
78°C (9.0 mmHg)
Authentic
Glass bottle
1.4910 to 1.494
MFCD00007461
1.55
VHCSBTPOPKFYIU-UHFFFAOYSA-N
2-chloroethanesulfonyl chloride
15385
95%

Chemical Identifiers

1622-32-8
163.02
VHCSBTPOPKFYIU-UHFFFAOYSA-N
15385
C(CCl)S(=O)(=O)Cl
C2H4Cl2O2S
MFCD00007461
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
2-chloroethanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-594-1

RTECSNumber : KI8060000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only