Learn More
2-Chloroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals
$68.25 - $68.25
Chemical Identifiers
CAS | 1622-32-8 |
---|---|
Molecular Formula | C2H4Cl2O2S |
Molecular Weight (g/mol) | 163.02 |
MDL Number | MFCD00007461 |
InChI Key | VHCSBTPOPKFYIU-UHFFFAOYSA-N |
Synonym | 2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride |
PubChem CID | 15385 |
IUPAC Name | 2-chloroethanesulfonyl chloride |
SMILES | C(CCl)S(=O)(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC404610050
|
Thermo Scientific Chemicals
404610050 |
5 g | Glass bottle |
Each for $68.25
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1622-32-8 | |
163.02 | |
VHCSBTPOPKFYIU-UHFFFAOYSA-N | |
15385 | |
C(CCl)S(=O)(=O)Cl |
C2H4Cl2O2S | |
MFCD00007461 | |
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride | |
2-chloroethanesulfonyl chloride |
Specifications
1622-32-8 | |
100.0 | |
1.5500g/mL | |
99°C | |
94% min. (ex Cl) (Argentometry) | |
C2H4Cl2O2S | |
ClCH2CH2SO2Cl | |
5 g | |
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride | |
C(CCl)S(=O)(=O)Cl | |
163.02 | |
163.02 | |
Liquid |
94.0 | |
Yellow to Brown | |
78°C (9.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4910 to 1.494 | |
MFCD00007461 | |
1.55 | |
VHCSBTPOPKFYIU-UHFFFAOYSA-N | |
2-chloroethanesulfonyl chloride | |
15385 | |
95% | |
2-Chloroethanesulfonyl chloride, 95% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 216-594-1
RTECSNumber : KI8060000
TSCA : TSCA
RUO – Research Use Only