Learn More
2-Chloroethanol, 99%, Thermo Scientific Chemicals
$43.53 - $311.86
Chemical Identifiers
CAS | 107-07-3 |
---|---|
Molecular Formula | C2H5ClO |
Molecular Weight (g/mol) | 80.511 |
MDL Number | MFCD00002829 |
InChI Key | SZIFAVKTNFCBPC-UHFFFAOYSA-N |
Synonym | ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol |
PubChem CID | 34 |
ChEBI | CHEBI:28200 |
IUPAC Name | 2-chloroethanol |
SMILES | C(CCl)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA36636AD
|
Thermo Scientific Chemicals
036636AD |
50 mL |
Each for $43.53
|
|
|||||
AA36636AK
|
Thermo Scientific Chemicals
036636AK |
250 mL |
Each for $91.21
|
|
|||||
AA36636K2
|
Thermo Scientific Chemicals
036636K2 |
1 L |
Each for $311.86
|
|
|||||
Description
2-Chloroethanol is used in the preparation of several dyes by the alkylation of aniline derivatives with chloroethanol. It is a building block in the production of pharmaceuticals and plasticizers. Also used for manufacture of thiodiglycol. It is a solvent for cellulose acetate and ethyl cellulose, textile printing dyes, in dewaxing, refining of rosin, extraction of pine lignin and the cleaning of machines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-07-3 | |
80.511 | |
SZIFAVKTNFCBPC-UHFFFAOYSA-N | |
34 | |
2-chloroethanol |
C2H5ClO | |
MFCD00002829 | |
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol | |
CHEBI:28200 | |
C(CCl)O |
Specifications
107-07-3 | |
Yellow | |
129°C | |
Ether-like | |
1.441 | |
MFCD00002829 | |
UN1135 | |
14,3793 | |
Fully miscible with water. | |
C(CCl)O | |
80.511 | |
CHEBI:28200 | |
99% | |
2-Chloroethanol |
-63°C | |
1.202 g/mL | |
55°C (131°F) | |
C2H5ClO | |
ClCH2CH2OH | |
50 mL | |
878139 | |
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol | |
SZIFAVKTNFCBPC-UHFFFAOYSA-N | |
2-chloroethanol | |
34 | |
80.51 | |
Liquid |
Safety and Handling
GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
H226-H300+H310+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYLENE CHLOROHYDRIN
EINECSNumber : 203-459-7
RTECSNumber : KK0875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only