Learn More
2-Chloroethyl phenyl sulfide, 98%, Thermo Scientific Chemicals
$68.11 - $211.84
Chemical Identifiers
CAS | 5535-49-9 |
---|---|
Molecular Formula | C8H9ClS |
Molecular Weight (g/mol) | 172.67 |
MDL Number | MFCD00013691 |
InChI Key | QDXIHHOPZFCEAP-UHFFFAOYSA-N |
Synonym | 2-chloroethyl phenyl sulfide, 2-chloroethyl phenyl sulfane, 2-chloroethyl sulfanyl benzene, 2-chloroethyl thio benzene, 2-chloroethylphenylsulfide, benzene, 2-chloroethyl thio, 2-phenylthio ethyl chloride, 2-phenylthioethylchloride, acmc-1axe6 |
PubChem CID | 21702 |
IUPAC Name | 2-chloroethylsulfanylbenzene |
SMILES | ClCCSC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2326506
|
Thermo Scientific Chemicals
B2326506 |
5 g |
Each for $68.11
|
|
||||||
AAB2326514
|
Thermo Scientific Chemicals
B2326514 |
25 g |
Each for $211.84
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5535-49-9 | |
172.67 | |
QDXIHHOPZFCEAP-UHFFFAOYSA-N | |
21702 | |
ClCCSC1=CC=CC=C1 |
C8H9ClS | |
MFCD00013691 | |
2-chloroethyl phenyl sulfide, 2-chloroethyl phenyl sulfane, 2-chloroethyl sulfanyl benzene, 2-chloroethyl thio benzene, 2-chloroethylphenylsulfide, benzene, 2-chloroethyl thio, 2-phenylthio ethyl chloride, 2-phenylthioethylchloride, acmc-1axe6 | |
2-chloroethylsulfanylbenzene |
Specifications
5535-49-9 | |
245°C | |
C8H9ClS | |
MFCD00013691 | |
UN2922 | |
2-chloroethyl phenyl sulfide, 2-chloroethyl phenyl sulfane, 2-chloroethyl sulfanyl benzene, 2-chloroethyl thio benzene, 2-chloroethylphenylsulfide, benzene, 2-chloroethyl thio, 2-phenylthio ethyl chloride, 2-phenylthioethylchloride, acmc-1axe6 | |
ClCCSC1=CC=CC=C1 | |
172.67 | |
172.68 | |
2-Chloroethyl phenyl sulfide |
1.178 | |
104°C (220°F) | |
1.583 | |
5 g | |
1907116 | |
QDXIHHOPZFCEAP-UHFFFAOYSA-N | |
2-chloroethylsulfanylbenzene | |
21702 | |
98% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 226-891-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only