missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Chloroethyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 215420250

 View more versions of this product

Catalog No. AC215420250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

(2-Chloroethyl)trimethylammonium chloride
999-81-5
100.0
White
98%
C5H13Cl2N
MFCD00011869
04, 134
chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
UHZZMRAGKVHANO-UHFFFAOYSA-M
2-chloroethyl(trimethyl)azanium;chloride
13836
98%
98%
97.5
239°C to 243°C
Authentic
Glass bottle
ClCH2CH2N(CH3)3Cl
25 g
15, 2104
Solubility in water: 740g/L (20°C)
C[N+](C)(C)CCCl.[Cl-]
158.07
158.07
Crystalline Powder or Crystals

Chemical Identifiers

999-81-5
158.07
UHZZMRAGKVHANO-UHFFFAOYSA-M
13836
C[N+](C)(C)CCCl.[Cl-]
C5H13Cl2N
MFCD00011869
chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
2-chloroethyl(trimethyl)azanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 213-666-4

RTECSNumber : BP5250000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only