Learn More
(2-Chloroethyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals
$110.38 - $264.74
Chemical Identifiers
CAS | 999-81-5 |
---|---|
Molecular Formula | C5H13Cl2N |
Molecular Weight (g/mol) | 158.07 |
MDL Number | MFCD00011869 |
InChI Key | UHZZMRAGKVHANO-UHFFFAOYSA-M |
Synonym | chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan |
PubChem CID | 13836 |
IUPAC Name | 2-chloroethyl(trimethyl)azanium;chloride |
SMILES | C[N+](C)(C)CCCl.[Cl-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC215420250
|
Thermo Scientific Chemicals
215420250 |
25 g |
Each for $110.38
|
|
|||||
AC215421000
|
Thermo Scientific Chemicals
215421000 |
100 g |
Each for $264.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
999-81-5 | |
158.07 | |
UHZZMRAGKVHANO-UHFFFAOYSA-M | |
13836 | |
C[N+](C)(C)CCCl.[Cl-] |
C5H13Cl2N | |
MFCD00011869 | |
chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan | |
2-chloroethyl(trimethyl)azanium;chloride |
Specifications
999-81-5 | |
100.0 | |
White | |
98% | |
C5H13Cl2N | |
MFCD00011869 | |
04, 134 | |
chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan | |
UHZZMRAGKVHANO-UHFFFAOYSA-M | |
2-chloroethyl(trimethyl)azanium;chloride | |
13836 | |
98% | |
(2-Chloroethyl)trimethylammonium chloride, 98% |
97.5 | |
239°C to 243°C | |
Authentic | |
Glass bottle | |
ClCH2CH2N(CH3)3Cl | |
25 g | |
15, 2104 | |
Solubility in water: 740g/L (20°C) | |
C[N+](C)(C)CCCl.[Cl-] | |
158.07 | |
158.07 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 213-666-4
RTECSNumber : BP5250000
TSCA : TSCA
RUO – Research Use Only