Learn More
2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1104814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloronicotinoyl chloride | |
38°C to 42°C | |
C6H3Cl2NO | |
25 g | |
119022 | |
2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride | |
ClC(=O)C1=CC=CN=C1Cl | |
176.00 | |
176 |
49609-84-9 | |
98°C to 100°C (2 mmHg) | |
MFCD00051677 | |
UN3261 | |
Moisture sensitive | |
RXTRRIFWCJEMEL-UHFFFAOYSA-N | |
2-chloropyridine-3-carbonyl chloride | |
2774541 | |
≥98% |
Chemical Identifiers
49609-84-9 | |
176.00 | |
RXTRRIFWCJEMEL-UHFFFAOYSA-N | |
2774541 | |
ClC(=O)C1=CC=CN=C1Cl |
C6H3Cl2NO | |
MFCD00051677 | |
2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride | |
2-chloropyridine-3-carbonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only