missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals

$483.93 - $1583.30

Chemical Identifiers

CAS 49609-84-9
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 176.00
MDL Number MFCD00051677
InChI Key RXTRRIFWCJEMEL-UHFFFAOYSA-N
Synonym 2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride
PubChem CID 2774541
IUPAC Name 2-chloropyridine-3-carbonyl chloride
SMILES ClC(=O)C1=CC=CN=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1104814
View Documents
Thermo Scientific Chemicals
A1104814
25 g
Each for $483.93
 
AAA1104822
View Documents
Thermo Scientific Chemicals
A1104822
100 g
Each for $1,583.30
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

49609-84-9
176.00
RXTRRIFWCJEMEL-UHFFFAOYSA-N
2774541
ClC(=O)C1=CC=CN=C1Cl
C6H3Cl2NO
MFCD00051677
2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride
2-chloropyridine-3-carbonyl chloride

Specifications

49609-84-9
98°C to 100°C (2 mmHg)
MFCD00051677
UN3261
Moisture sensitive
RXTRRIFWCJEMEL-UHFFFAOYSA-N
2-chloropyridine-3-carbonyl chloride
2774541
≥98%
38°C to 42°C
C6H3Cl2NO
25 g
119022
2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride
ClC(=O)C1=CC=CN=C1Cl
176.00
176
2-Chloronicotinoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only