Learn More
2-Chlorophenylboronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 358990050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Chlorophenylboronic acid | |
96.0 | |
Authentic | |
Glass bottle | |
MFCD00674012 | |
2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl | |
RRCMGJCFMJBHQC-UHFFFAOYSA-N | |
(2-chlorophenyl)boronic acid | |
2734322 | |
97% |
3900-89-8 | |
100.0 | |
96% min. (HPLC) | |
C6H6BClO2 | |
5g | |
Solubility in water: insoluble. Other solubilities: soluble in ether,thf,dmso,dmf,methanol | |
B(C1=CC=CC=C1Cl)(O)O | |
156.38 | |
156.38 |
Chemical Identifiers
3900-89-8 | |
156.38 | |
RRCMGJCFMJBHQC-UHFFFAOYSA-N | |
2734322 | |
B(C1=CC=CC=C1Cl)(O)O |
C6H6BClO2 | |
MFCD00674012 | |
2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl | |
(2-chlorophenyl)boronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO â Research Use Only